Dishes, crystallising, quartz
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8.653.001 8.653.002 8.653.003
WITG8.653.001EA 0 GBP
WITG8.653.001 WITG8.653.002 WITG8.653.003
Dishes, crystallising, quartz
Trays/Dishes/Pots Crystallising Dishes
Crystallising dish made of quartz-silica, with spout.
  • Extremely low expansion
  • Best possible elastricity and thermal shock resistance
  • High transformation and softening point temperature
  • Low heat conductivity
  • Low electrical conductivity
For a wide range of applications.