Woulff bottles
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.801.001 5.801.500
WITG5.801.001EA 182 GBP
WITG5.801.001 WITG5.801.500
Woulff bottles
Bottles Woulff Bottles
Made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion gives relatively high resistance to temperature changes
With two side necks GL 18, center neck GL 25, complete with screw caps.