Spinner flasks, BIOGEN
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5.529.005 5.529.007 5.529.008 5.529.001 5.529.002 5.529.003 5.529.004 5.529.006 5.529.009
WITG5.529.005EA 883 GBP
WITG5.529.005 WITG5.529.007 WITG5.529.008 WITG5.529.001 WITG5.529.002 WITG5.529.003 WITG5.529.004 WITG5.529.006 WITG5.529.009
Spinner flasks, BIOGEN
Flasks Spinner Flasks
Borosilicate glass with side arms
The fluid dynamic impeller assemblies to assure optimal lift of microcarrier beads at very low mixing speeds, the domed vessel bottom prevents microcarrier accumulations, positive-locking compression seal provides adjustable impeller assembly height, constructed of non-cytotoxic material, flasks inside surface siliconized on request.