Centrifuge tubes, conical bottom
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4.945.005
WITG4.945.005EA 26.7 GBP
WITG4.945.005
Centrifuge tubes, conical bottom
Tubes Centrifuge Tubes
DURAN®, borosilicate glass.
  • Heavy wall
  • Ungraduated
  • Very good chemical resistance
  • Very good temperature resistance
Conical bottom, without rim