Volumetric flasks, trapezoidal shape, class A, brown or blue markings
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3.669.000 3.669.001 3.669.050 3.669.000.BR 3.669.001.BR 3.669.500 3.669.002.BL
WITG3.669.000EA 37.8 GBP
WITG3.669.000 WITG3.669.001 WITG3.669.050 WITG3.669.000.BR WITG3.669.001.BR WITG3.669.500 WITG3.669.002.BL
Volumetric flasks, trapezoidal shape, class A, brown or blue markings
Flasks Volumetric Flasks
Borosilicate glass 3.3 with PE stopper.
  • DE-M marked according to the German calibration law
  • Inclusive batch certificate
  • Brown or blue markings
Certifications: DIN EN ISO 1042