Separating funnels, spherical
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2.620.050
WITG2.620.050EA 164 GBP
WITG2.620.050
Separating funnels, spherical
Funnels Separating Funnels
Separatory funnel, globe (ball) shaped, made of borosilicate glass 3.3, with detachable PTFE-stopcock and PE stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance