Destilling receivers acc. to Bernhauer
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.932.001 1.934.001 1.930.001 1.936.001
WITG1.932.001EA 127 GBP
WITG1.932.001 WITG1.934.001 WITG1.930.001 WITG1.936.001
Destilling receivers acc. to Bernhauer
Distillation Components Distillation Receivers
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes