Adapters, delivery, straight
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.900.019 1.900.024 1.900.029
WITG1.900.019EA 26.6 GBP
WITG1.900.019 WITG1.900.024 WITG1.900.029
Adapters, delivery, straight
Distillation Components Distillation Adapters
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes