Funnels, with standard ground joint cone
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.650.014 1.650.029 1.650.129 1.650.130 1.650.150 1.650.019
WITG1.650.014EA 14.9 GBP
WITG1.650.014 WITG1.650.029 WITG1.650.129 WITG1.650.130 WITG1.650.150 WITG1.650.019
Funnels, with standard ground joint cone
Funnels Filtration Funnels Funnels with Conical Joints
Funnels, made of boroslicate glass, with standard ground joint cone.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for transferring liquids or powders to glassware with standard ground joints.