Sulfonation flasks, with three necks
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0.851.020 0.851.101 0.851.024 0.851.074 0.851.150 0.851.250
WITG0.851.020EA 154 GBP
WITG0.851.020 WITG0.851.101 WITG0.851.024 WITG0.851.074 WITG0.851.150 WITG0.851.250
Sulfonation flasks, with three necks
Flasks Sulfonation Flasks
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one parallel and two angled side necks.