Gaskets for screw caps
Supplier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0.180.012
WITG0.180.012EA 17.2 GBP
WITG0.180.012
Gaskets for screw caps
Seals Gaskets
Silicone with PTFE washers
  • Temperature resistant from −200 to +250 °C
  • Very good chemical resistance
  • Easy handling
For threaded connection caps with hole and GL thread.