Roller culture apparatus, Wheaton® R2P™ 2.0
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WRBPF8110-C WRBPF5110-C WRBPF5111-C WRBPF8111-C WRBMR5070-C WRBMR5031-C WRBMR5010-B WRBMR5030-J WRTPF5110-B WRTPF8110-F WRTPF8110-D WRTPF8110-C WRTPF8110-B WRTPF8110-A WRTPF5110-J WRTPF5110-F WRBPF5110-B WRBMR5030-F WRBMR5010-A WRBPF5110-A WRBMR5090-J WRBMR5090-F WRBMR5090-D WRBMR5090-C WRBMR5090-B WRBMR5090-A WRBPF5110-D WRBPF5110-F WRBMR5030-D WRBMR5030-C WRBMR5030-B WRBMR5030-A WRBMR5010-J WRBMR5010-F WRBMR5010-D WRBMR5010-C WRTPF5110-C WRTPF8110-J WRTPF5110-A WRBPR5030-J WRBPR5030-F WRBPR5030-D WRBPR5030-C WRBPR5030-B WRBPR5030-A WRBPF5110-J WRTPF5110-D WSBMR5040-C WSBMR5040-D WSBMR5020-D WSTPF8110-C WSTPF8111-C WSBPF5110-C WRBMR5091-C WSTPF5111-C WSBMR5010-C WSBMR5090-C WRBPR5050-C WSBMR5011-D WSBMR5010-D WRBMR5071-C WRTMR5071-C
WHEAWRBPF8110-CEA 39490 GBP
WHEAWRBPF8110-C 392-0349 WHEAWRBPF5111-C WHEAWRBPF8111-C 392-0454 WHEAWRBMR5031-C WHEAWRBMR5010-B WHEAWRBMR5030-J WHEAWRTPF5110-B WHEAWRTPF8110-F 392-0356 392-0355 WHEAWRTPF8110-B WHEAWRTPF8110-A WHEAWRTPF5110-J WHEAWRTPF5110-F WHEAWRBPF5110-B WHEAWRBMR5030-F WHEAWRBMR5010-A WHEAWRBPF5110-A WHEAWRBMR5090-J WHEAWRBMR5090-F 392-0456 392-0455 WHEAWRBMR5090-B WHEAWRBMR5090-A 392-0350 WHEAWRBPF5110-F 392-0453 392-0452 WHEAWRBMR5030-B WHEAWRBMR5030-A WHEAWRBMR5010-J WHEAWRBMR5010-F WHEAWRBMR5010-D WHEAWRBMR5010-C 392-0353 WHEAWRTPF8110-J WHEAWRTPF5110-A WHEAWRBPR5030-J WHEAWRBPR5030-F 392-0352 392-0351 WHEAWRBPR5030-B WHEAWRBPR5030-A WHEAWRBPF5110-J 392-0354 WHEAWSBMR5040-C WHEAWSBMR5040-D WHEAWSBMR5020-D WHEAWSTPF8110-C WHEAWSTPF8111-C WHEAWSBPF5110-C WHEAWRBMR5091-C WHEAWSTPF5111-C WHEAWSBMR5010-C 392-0458 WHEAWRBPR5050-C WHEAWSBMR5011-D WHEAWSBMR5010-D WHEAWRBMR5071-C WHEAWRTMR5071-C
Roller culture apparatus, Wheaton® R2P™ 2.0
Roller Culture Apparatus
The Wheaton® R2P™ 2.0 roller culture apparatus is designed to allow for the development and growth of cultured cells from research to production quantities without having to change equipment and revalidate protocols. The R2P™ 2.0 system utilises computerised measurement of the actual bottle speed and allows networking for up to 255 units. Applications include, Viruses for vaccines or related studies, proteins, enzymes, antibodies, live cell assays.
  • Robust touch screen interface that is easy to view from a distance
  • Readily recognised icons for intuitive
More Product Information