Distillation glassware set with standard taper 19/22 Clear-Seal® joints, Micro Kit®, WHEATON®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
109933 123753 168353 964453 923769 773900 690170
WHEA109933EA 116 GBP
WHEA109933 WHEA123753 WHEA168353 WHEA964453 WHEA923769 WHEA773900 WHEA690170
Distillation glassware set with standard taper 19/22 Clear-Seal® joints, Micro Kit®, WHEATON®
Distillation Components Distillation Sets
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Superior in vacuum and radioactive applications
  • Smooth surfaces resist contamination
  • Cleaning is simple and thorough
  • Interchangeable with ground joints
Apparatus for a wide range of organic chemistry experiments at a lower price than purchasing the components separately.