Qualitative filter papers, standard grades, grade 597 Optima, Whatman™
Supplier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
989410108 989410109 989410107 989410106 989410105 989410110 98949613 989410104 989410103 98949329 989410102 98949552 989410101
WHAT989410108EA 51.8 GBP
WHAT989410108 WHAT989410109 WHAT989410107 WHAT989410106 WHAT989410105 WHAT989410110 WHAT98949613 WHAT989410104 WHAT989410103 WHAT98949329 WHAT989410102 WHAT98949552 WHAT989410101
Qualitative filter papers, standard grades, grade 597 Optima, Whatman™
Filters Filter Papers Qualitative Filter Paper
Standard Whatman grade filter paper for a variety of routine applications in food and beverage industries. Well-suited for determination of fat content in food or removal of carbon dioxide and turbidity from beverages such as beer.
  • Available in sheets and circles
  • Medium fast filtration rate
  • Minimum 90% cellulose content
  • Technical and scientific support
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.