Quartz microfibre filters for air sampling, grade QM-A, Whatman™
Supplier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1851-025 1851-037 1851-047 1851-050 1851-055 1851-085 1851-865 1851-150 1851-032 1851-082 1851-090 1851-101 1851-110 1851-118 1851-8866 1851-045 1851-037_L
512-0430EA 200 GBP
512-0430 516-2608 516-2611 516-5116 516-0199 516-5119 512-0431 516-5122 513-0112 512-0671 513-0086 513-0113 516-5120 516-5121 516-5123 WHAT1851-045 WHAT1851-037_L
Quartz microfibre filters for air sampling, grade QM-A, Whatman™
Filters Glass Fibre Filters
These high purity quartz microfibre filters are used in many air sampling filter applications, including sampling of acidic gases, stacks, flues and aerosols. They are suitable for PM-10 sampling and analysis.
  • Binder-free
  • High loading capacity
Because of the low level of alkaline earth metals, 'artifact' products of sulfates and nitrates (from SO₂ and NO₂) are virtually eliminated. High-purity quartz microfiber filters are therefore applicable for particulate monitoring and heavy metal analysis.