Cellulose chromatography and blotting paper roll, Whatman™
Supplier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
10539028
WHAT10539028EA 120 GBP
WHAT10539028
Cellulose chromatography and blotting paper roll, Whatman™
Filters Filter Papers Chromatography Paper
Cellulose Chromatography and Blotting Paper Roll contains cotton linter filter sheets for electrophoresis and amino acid chromatography.
It is also ideal for use with gelatinous samples.