Test tube racks, anodised aluminium
Supplier: WESBART
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SP2384 SP3012 DP1120 DP1148 DP1450 DP14144 DP1820 DP1830 DP1840 DP1850 DP18100 DP1848 DP18120 DP2030 DP2050 DP2036
WESBSP2384EA 69.2 GBP
WESBSP2384 WESBSP3012 WESBDP1120 WESBDP1148 WESBDP1450 WESBDP14144 WESBDP1820 WESBDP1830 WESBDP1840 WESBDP1850 WESBDP18100 WESBDP1848 WESBDP18120 WESBDP2030 WESBDP2050 WESBDP2036
Test tube racks, anodised aluminium
Racks Tube Racks
These test tube racks are manufactured from 1 mm anodised aluminium designed to be durable in any laboratory.
  • Lightweight
  • Sturdy
  • Excellent temperature resistance
Test Tube racks are an effective way of storing Falcon/ Universal tubes, Bijous, cryogenic vials or any size glass bottles.