Disinfectant tablets, PRESEPT™
Supplier: WESTERN LABORATORY SERVICE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SPR25
WELASPR25EA 138 GBP
WELASPR25
Disinfectant tablets, PRESEPT™
Cleaning Agents Disinfectants
These 2,5 g tablets contain sodium dichloroisocyanurate (NaDCC) in an effervescent base. The aqueous solution is rapid acting, and is effective against bacteria, spores, fungi, algae, protozoa and viruses, (including AIDS virus) and is resistant to inactivation by organic soilage.
  • Activity across the complete biocidal spectrum
  • Suitable for cleaning and disinfecting work surfaces, utensils and linen etc
  • Convenient tablet format, more stable and better biocidal activity than hypochlorites
  • Solution is compatible with anionic and non ionic detergents