Preparative columns, µBondapak® C18
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WAT085721
WATEWAT085721EA 467 GBP
WATEWAT085721
Preparative columns, µBondapak® C18
Chromatography Columns
µBondapak® C18 columns are general purpose, silica-based, reversed phase C18 columns that are based on 10 µm particle technology.