Glycan BEH amide columns, ACQUITY UPLC™
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186004742 186004741 186004740
WATE186004742EA 866 GBP
WATE186004742 WATE186004741 WATE186004740
Glycan BEH amide columns, ACQUITY UPLC™
Chromatography Columns
The ACQUITY UPLC BEH glycan separation technology column, also known as BEH Glycan, is optimized for use with ACQUITY UPLC systems with fluorescence detection (FLR), separating the released glycans of biopharmaceuticals as their 2-aminobenzamide (2-AB) derivatives.
  • Suitable for UPLC and UHPLC systems