Analytical HPLC columns, XBridge™ BEH C18
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186003031 186003022 186002978 186003034 186003027 186003110 186003111
WATE186003031EA 666 GBP
WATE186003031 WATE186003022 WATE186002978 WATE186003034 WATE186003027 WATE186003110 WATE186003111
Analytical HPLC columns, XBridge™ BEH C18
Chromatography Columns
XBridge™ BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12). The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.