UPLC columns, ACQUITY UPLC™ BEH Shield RP 18
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186002853 186002854
WATE186002853EA 766 GBP
WATE186002853 WATE186002854
UPLC columns, ACQUITY UPLC™ BEH Shield RP 18
Chromatography Columns
These columns complement the ACQUITY BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.