Analytical HPLC columns, SunFire™ C8
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186002731 186002730
WATE186002731EA 693 GBP
WATE186002731 WATE186002730
Analytical HPLC columns, SunFire™ C8
Chromatography Columns
SunFire™ columns provide symmetrical peak shapes for improved resolution. With new bonding and endcapping technologies, they offer great efficiency for acid, neutral, and basic compounds at low and intermediate pH ranges. They are compatible with mass spectrometry applications, providing sharp peaks, high sensitivity, large peak capacities and very low bleed.