Reversed phase HPLC columns, µBondapak® C18
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WAT086684 WAT088070 186000925 WAT044480
WATEWAT086684EA 747 GBP
WATEWAT086684 WATEWAT088070 WATE186000925 WATEWAT044480
Reversed phase HPLC columns, µBondapak® C18
Chromatography Columns
µBondapak® C18 columns are general purpose, silica-based, reversed phase C18 columns that are based on 10 µm particle technology.