Analytical HPLC columns, XTerra® RP8
Supplier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186000439
WATE186000439EA 693 GBP
WATE186000439
Analytical HPLC columns, XTerra® RP8
Chromatography Columns
XTerra® RP8 columns are general purpose, hybrid-based, reversed phase C8 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Order Now
SPECIFICATIONS
Environmentally Preferable