Laboratory blenders
Supplier: WARING BLENDOR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CB15E CB15TE
WARICB15EEA 10670 GBP
WARICB15E WARICB15TE
Laboratory blenders
Homogenisers Standard Laboratory Blenders
The CB15 series blenders are designed specifically for processing large quantities of free flowing material with a high liquid content. This range of 3-speed blenders has an integral pulse button and is fitted with a powerful 3,75 hp motor encased within a die-cast epoxy coated motor housing.
  • Easy to move and fitted with full traction rubber feet for greater stability
  • Easy-to-clean electronic keypad
  • Slow Start™ blending mechanism
  • Stainless steel container jar with two handles
  • Smaller 2, 1 litre and mini-containers can be used with the aid of the AD
More Product Information