Nitric acid 0.2 mol/l (0.2 N)

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Nitric acid solution

VWRS5550.5000EA 0 GBP
VWRS5550.5000
Nitric acid 0.2 mol/l (0.2 N)
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR