VWR® Bottles, Wide Neck, Square, with Screw Caps
Supplier: LAB3 OWN PRODUCTION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VWRLSTL101EA 73.9 GBP
VWRLSTL101 VWRLSTL102 VWRLSTL13 VWRLSTL100 VWRLSTL101.1
VWR® Bottles, Wide Neck, Square, with Screw Caps
Bottles Plastic Bottles
PET, clear, with screw cap.
  • Good chemical resistance
  • With or without label available
Ideal for packaging, storing and transporting liquids, pastes and powders. Wide mouth for easy filling, emptying and cleaning.