VWR® Water Sampling Bottles
Supplier: LAB3 OWN PRODUCTION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VWRL080257EA 82.5 GBP
VWRL080257 VWRL080259 VWRL080260 VWRL080269 VWRL080274 VWRL080275
VWR® Water Sampling Bottles
Bottles Water Sampling Bottles
Soda-Lime glass, round, clear or amber.
  • Rigid and robust
  • Leak proof with correct cap
  • PTFE lined caps, liners fixed in so do not fall out
  • Dosed with 18 mg/l sodium thiosulphate (18%)
  • Expiry 12 months from manufacture
Ideal for water and general testing, filling and storage. Amber coloured for light sensitive materials.
Certifications: Standards of British ,European (2002 ) and U.S. (25) Pharmacopoeia
Amber to above and BS1679
Meet regulations 2005 (UK) and no.1935/2004 (EC) DMF No’s 10500/10501 (U.S.)articles in contact with food