VWR® Bottles, Water Sampling
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
331-0278EA 145 GBP
331-0278 331-0279 331-0280 331-0281 331-0282 331-0286 VWRI331-0287.1 VWRI331-0288.1 VWRI331-0283.1 VWRI331-0278.1 VWRI331-0279.1 VWRI331-0280.1 VWRI331-0281.1 VWRI331-0282.1 VWRI331-0286.1 VWRI331-0287_U VWRI331-0288_U
VWR® Bottles, Water Sampling
Bottles Water Sampling Bottles
PET, virgin, food grade with PP caps.
  • Irradiated - sterile
  • Different coloured caps for easy indentification of test
  • Tamper-evident closure to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Tamper-evident screw caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Square bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.
Order Now
SPECIFICATIONS
Environmentally Preferable