VWR®, Water Sampling Bottles, Sterile
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
331-0256EA 363 GBP
331-0256 331-0267 331-0268 331-0269 331-0270 331-0271 331-0272 331-0273 331-0274 331-0275 331-0276 331-0277 VWRI331-0256.1 VWRI331-0267.1 VWRI331-0268.1 VWRI331-0269.1 VWRI331-0270.1 VWRI331-0271.1 VWRI331-0272.1 VWRI331-0273.1 VWRI331-0274.1 VWRI331-0275.1 VWRI331-0276.1 VWRI331-0277.1
VWR®, Water Sampling Bottles, Sterile
Bottles Water Sampling Bottles
PET, virgin, food grade with PP caps.
  • Different coloured caps for easy indentification of test
  • Tamper-evident strip to ensure sampling integrity
  • Full traceability, batch number and individual bottle number on each label
  • Fully recyclable
  • Cap diameter: 32 mm
Lightweight, durable bottles. Caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Rectangular bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.
Red cap: For chlorinated water dosed with 20More Product Information