VWR® Media Bottles, Square, PETG

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

VWRI210-000057EA 162 GBP
VWRI210-000057 VWRI210-000071 VWRI210-000070 VWRI210-000068 VWRI210-000056 VWRI210-000082 VWRI210-000066 VWRI210-000063 VWRI210-000073 VWRI210-000084 VWRI210-000064 VWRI210-000081 VWRI210-000069 VWRI210-000058 VWRI210-000080 VWRI210-000062 VWRI210-000065 VWRI210-000075 VWRI210-000059 VWRI210-000060 VWRI210-000074 VWRI210-000067 VWRI210-000079 VWRI210-000072 VWRI210-000076 VWRI210-000083 VWRI210-000061 VWRI210-000078 VWRI210-000077 VWRI210-000055
VWR® Media Bottles, Square, PETG
Bottles Cell Culture Bottles
Square PETG bottles are ideal for storage of media, laboratory reagents and biological buffers up to −70 °C. Eliminate washing, depyrogenation and autoclaving steps with sterile and non-pyrogenic.

  • Made from 100% virgin PETG material, durable and break resistant
  • Heat-shrink band around closure and neck for tamper-evident seal
  • Leakproof bottles and HDPE closures provided
  • Leakproof sealing with integrated lip seal feature
  • Sterile and non-pyrogenic, SAL 10⁻⁶
  • Low extractables
  • Transparency and reduced permeability
  • Arranged in shrink-wrapped trays and polybag for
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR