Klebsiella pneumoniae WDCM 00097 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VT000971-10EA VT000975-10EA
VT000971-10EAEA 97.1 GBP
VT000971-10EA VT000975-10EA
Klebsiella pneumoniae WDCM 00097 Vitroids™, Certified Reference Materials, Suitable for Microbiology, Supelco®
Standards Biological Standards
Derived from CECT® strains.
  • Origin Strain No. CECT® 143
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: Vitroids is a trademark of Sigma-Aldrich Co. LLC