Wash bottles, narrow or wide neck
Supplier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
94993 93793 95093 93993 95193 94193
VITL94993EA 10.3 GBP
VITL94993 VITL93793 VITL95093 VITL93993 VITL95193 VITL94193
Wash bottles, narrow or wide neck
Bottles Wash Bottles
PP, translucent, screw cap and spray tube made from PP.
  • Excellent chemical resistance
  • Excellent temperature resistance
Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.