Measuring beakers with handle
Supplier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
44291 44391 44091 44191 44491
VITL44291EA 13.5 GBP
VITL44291 VITL44391 VITL44091 VITL44191 VITL44491
Measuring beakers with handle
Beakers
SAN, crystal clear.
  • Temperature resistance up to 70 °C
  • With a raised scale
  • Stable, easy-grip handle
Ideal for general laboratory use and a wide variety of other applications. With embossed scale.