Single channel pipettes, fixed volume, MLA™ Micro D-Tipper™
Supplier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1802 1803 1804 1805 1806 1807 1808 1809 1810
VIST1802EA 448 GBP
VIST1802 VIST1803 VIST1804 VIST1805 VIST1806 VIST1807 VIST1808 VIST1809 VIST1810
Single channel pipettes, fixed volume, MLA™ Micro D-Tipper™
Pipettes Micropipettes
Micro D-Tipper™ pipettes are general purpose pipettes for sampling and dispensing liquids when accuracy, precision and convenience are required. The Micro D-Tipper pipettes are available in nine volumes ranging from 2 to 10 µl and use a standard MLA small tip.
  • Piston stroke, air displacement instruments with tip ejection
  • Two stroke pipette with overblow, enhancing the accuracy and precision at volumes below 10 µl
  • Factory calibrated to dispense the volume engraved on the pipette
  • Calibration can be verified and easy in- lab adjustment is possible
Order Now
SPECIFICATIONS
Environmentally Preferable