Single channel pipettes, fixed volume, MLA D-Tipper™
Supplier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1051C 1055C 1060C 1152C 1052C 1056C 1053C 1057C 1058C 1153C 1141C 1054C 1059C 1159C 1162C 1148C 1149C 1160C 1163C 1142C 1143C 1144C 1056CLT 1146C 1147C 1150C 1151C 1154C 1157C
VIST1051CEA 362 GBP
VIST1051C VIST1055C VIST1060C VIST1152C VIST1052C VIST1056C VIST1053C VIST1057C VIST1058C VIST1153C VIST1141C VIST1054C VIST1059C VIST1159C VIST1162C VIST1148C VIST1149C VIST1160C VIST1163C VIST1142C VIST1143C VIST1144C VIST1056CLT VIST1146C VIST1147C VIST1150C VIST1151C VIST1154C VIST1157C
Single channel pipettes, fixed volume, MLA D-Tipper™
Pipettes Micropipettes
These superior performance MLA pipettes are ideal for clinical work.
  • Constructed with precision engineered aluminum and stainless steel
  • Light plunger forces for easy operation with thumb/ forefinger
  • Colour-coded body for easy volume identification
  • In-lab calibration adjustment
  • Single-stroke 'to deliver' operation
  • Built in tip-ejector
  • Glove guard