Single channel macro pipettes, fixed volume, MLA
Supplier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1065 1066 1067 1068 1069 1070 1076 1077 1078 1079
VIST1065EA 448 GBP
VIST1065 VIST1066 VIST1067 VIST1068 VIST1069 VIST1070 VIST1076 VIST1077 VIST1078 VIST1079
Single channel macro pipettes, fixed volume, MLA
Pipettes Micropipettes
These macro volume pipettes are available in 10 fixed volumes ranging from 1 to 10 ml.
  • Superior performance
  • Single-stroke 'to deliver' operation
  • Constructed with precision engineered aluminum and stainless steel
  • Colour-coded body for easy volume identification
  • Light plunger forces for easy operation with thumb/ forefinger