MultiDuster mopping systems
Supplier: FHP VILEDA SCS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
151612 151719
VILE151612EA 30.4 GBP
VILE151612 VILE151719
MultiDuster mopping systems
Mops
MultiDuster has a unique mechanism that brings significant benefits like hygienic cleaning of high and inaccessible areas.
  • Easy to wipe clean
  • Doesn’t harbour bacteria
  • Fits to handle in all bent positions
  • Double sided sleeves made of microfibre
  • Returns to straight position
  • Fits telescopic handles
MultiDuster Maxi easily cleans pipes and ledges up to 5 m high. For low areas the MultiDuster Mini is the perfect hand-held solution. Both Maxi and Mini have slim designs, for easy cleaning behind tight spaces such as radiators, between glass partitions,More Product Information