Replacement tubes
Supplier: Vilber Lourmat
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
907500631 907501521 907510811 907510831 907511521 907510821 907520411 907520611 907520631 907511511 907511531 907520831 907521511 907500611 907500811 907500821 907500831 907501511 907501531 907520811 907520821 907521521 907521531
VILB907500631EA 0 GBP
VILB907500631 VILB907501521 VILB907510811 VILB907510831 VILB907511521 VILB907510821 VILB907520411 VILB907520611 VILB907520631 VILB907511511 VILB907511531 VILB907520831 VILB907521511 VILB907500611 VILB907500811 VILB907500821 VILB907500831 VILB907501511 VILB907501531 VILB907520811 VILB907520821 VILB907521521 VILB907521531
Replacement tubes
Lights UV Lights, Lamps and Bulbs
Replacement tubes for transilluminators and UV lamps.
  • High UV irradiation and constant spectra
  • Longer shelf-life for thousands of irradiation hours
  • Unique electrode allowing for the highest reliability
Large output and wavelength ranges available.