UV transilluminators, multibrand
Supplier: Vilber Lourmat
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
216120151 216120171 216126151 216126161 216126141 216126171
VILB216120151EA 0 GBP
VILB216120151 VILB216120171 VILB216126151 VILB216126161 VILB216126141 VILB216126171
UV transilluminators, multibrand
Transilluminators UV Transilluminators
This UV transilluminator accommodates a wide range of applications requiring different wavelengths for both visualisation and documentation.
  • UV protection shield
  • Two UV wavelengths in one single transilluminator
  • Super-Bright filter to stop the visible light from UV tubes for an unbeatable viewing field
15-Watt W×D×H: 470×330×150 mm