UV transilluminators, high/low intensity
Supplier: Vilber Lourmat
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
213115011 213115021 213120111 213120121 213120131 213126101 213126111 213126121
VILB213115011EA 0 GBP
VILB213115011 VILB213115021 VILB213120111 VILB213120121 VILB213120131 VILB213126101 VILB213126111 VILB213126121
UV transilluminators, high/low intensity
Transilluminators UV Transilluminators
The UV transilluminators is a compact model equipped with the Super Bright filter, which stops the visible light emitted by the UV tubes affording an unbeatable visualisation quality.
  • Adjustable UV safety screen can be positioned to suit the operator's viewing angle against exposure to harmful UV rays
  • Intensity selector for short gel visualisation or longer preparative work
  • UV Master® technology incorporating Ondulex® reflector for higher UV output
W×D×H: 340×285×146
Power: 100 to 240 V, 50/60 Hz