DEC-QUAT 200C, disinfectant
Supplier: VELTEK ASSOCIATES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DQ200C0716Z01E
VELTDQ200C0716Z01EEA 552 GBP
VELTDQ200C0716Z01E
DEC-QUAT 200C, disinfectant
Cleaning Agents Disinfectants
DEC-QUAT 200C is a disinfectant, non-food contact sanitizer, cleaner, mildewstat, fungicide, virucide, and deodorizer. DEC-QUAT 200C is a fifth generation quaternary ammonium solution.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Filtered at 0.2 microns
  • One-step disinfectant for use in chemical cleaning rotation cycles
  • Phosphate-free
DEC-QUAT 200C is effective as a broad spectrum hospital disinfectant, fungicide, deodorizer, hard surface disinfectant, and non-food contact sanitizer effective against a broad spectrum of bacteria and viruses. DEC-QUAT 2More Product Information