Vortex mixers, RX3 and ZX3
Supplier: VELP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F202A0176 F202A0171 SA202A0176
444-2000EA 229 GBP
444-2000 444-0915 VELPSA202A0176
Vortex mixers, RX3 and ZX3
Shakers Vortex Shakers
Vortex mixers with epoxy painted aluminium die-cast housing that provides good chemical resistance. Four feet give stability on the bench. Choice of two models.
  • RX3 which operates in touch mode at a fixed vibration speed of 3000 min⁻¹
  • ZX3 features two modes - manual or continuous, speed can be adjusted from 0 to 3000 min⁻¹
  • The maximum vibration speed may be lower when adding on additional accessories (max. 800 min⁻¹), as high vibration speed may cause the sample to spill or the tube to fly out
IP Protection Class according to DIN EN 60259: IP 20