VARIO® PC 3010 NT VARIO select Chemistry Pumping Unit
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25744852
VACU25744852EA 11400 GBP
VACU25744852
VARIO® PC 3010 NT VARIO select Chemistry Pumping Unit
Pumps Vacuum Diaphragm Pumps
Chemistry diaphragm pumps from VACUUBRAND for aggressive gases and vapors are available in the basic version, with two-point or VARIO® control. Their construction with fluoropolymers makes them very resistant to chemical vapors from inlet to exhaust.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Ideal for evaporation of large amounts of high-boiling solvents even at low temperatures
More Product Information