Rotary Vane Pumps, RE 6
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20797162
VACU20797162EA 1910 GBP
VACU20797162
Rotary Vane Pumps, RE 6
Pumps Vacuum Rotary Vane Pumps
Rotary vane pumps from VACUUBRAND offer great performance in a compact design. Even with the gas ballast open, they enable a very good ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to optimised lubrication circuit and large usable oil volume.
  • High flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance (40 mbar) due to efficient gas ballast
  • Very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off; external anti-suckback valve not needed
  • Large oil volume: Long
More Product Information