Indium sealing for small flange connections
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20660150 20660151 20660152
VACU20660150EA 75.2 GBP
VACU20660150 VACU20660151 VACU20660152
Indium sealing for small flange connections
Seals Sealing Rings
Indium sealing for KF connections with stainless steel/aluminum centering rings (inside/outside) for small flange.