Centring and sealing rings for small flange connections
Supplier: VACUUBRAND
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20660140 20660141 20660143
VACU20660140EA 11.9 GBP
VACU20660140 VACU20660141 VACU20660143
Centring and sealing rings for small flange connections
Seals Sealing Rings
These centring and sealing rings for small stainless steel flange KF connections. With aluminium sealing ring.