Filtered UV lamps, UVI Light
Supplier: UVITEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LF106L LF106S LF206LM LF215L LF215M
UVLTLF106LEA 0 GBP
UVLTLF106L UVLTLF106S UVLTLF206LM UVLTLF215L UVLTLF215M
Filtered UV lamps, UVI Light
Lights UV Lights, Lamps and Bulbs
These ultraviolet lamps include low power handheld models to high intensity units. There are filtered lamps (three wavelengths) for use in fluorescent techniques and unfiltered lamps in both long and short (germicidal) wavelengths. The user friendly shape permits lamps to be held easily with gloved hands. These highly efficient, well designed lamps are produced in three wavelengths (254, 312 and 365 nm) and dual wavelength combinations.
  • Short wavelength (254 nm) models for fluorescence techniques, fluorochemistry, mineralogy, photopolymerisation, mutation
More Product Information