Safety helmets, uvex airwing B-S
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9762.521 9762.421
UVEX9762.521EA 24.6 GBP
UVEX9762.521 UVEX9762.421
Safety helmets, uvex airwing B-S
Head Protection Safety Helmets
Polyethylene helmet shell with three variable air vents and conventional suspension harness for a comfortable fit.
  • Shorter brim for a wider field of upward vision
  • Provides protection even in extremely low temperatures
Size: 510 to 610 mm
Certifications: EN 397